Would you like to log in again?
Cathepsin H Activity Assay Kit, BioVision
Supplier: BioVision
K145-100
10013-600
Cathepsin H Activity Assay Kit, BioVision
  • Fluorescence (Ex/Em 400/505)
  • Mammalian
  • 100 assays
  • Detects alternative proteolytic enzymes such as the lysosomal cathepsin proteases that can initiate or propagate proapoptotic signals
Apoptosis can be mediated by mechanisms other than the traditional caspase-mediated cleavage cascade. There is growing recognition that alternative proteolytic enzymes such as the lysosomal cathepsin proteases may initiate or propagate proapoptotic signals. Cathepsins are lysosomal enzymes that are also used as sensitive markers in various toxicological investigations. The Cathepsin-H Activity Assay kit is a fluorescence-based assay that utilizes the preferred cathepsin-H substrate Arginine labeled with AFC (amino-4-trifluoromethyl coumarin). Cell lysates or other samples that contain cathepsin-H will cleave the synthetic substrate R-AFC to release free AFC. The released AFC can easily be quantified using a fluorometer or fluorescence plate reader.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty