Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

High Pressure In-Line Filter Holder, Stainless Steel, Advantec MFS

Supplier: ADVANTEC MFS MS
304900
High-quality and durable, this 47 mm stainless steel in-line pressure holder can operate at high pressures up to 1420 psi (9.8 MPa).

Standard Viton O-rings are compatible with many aggressive liquids and gases. Type 304ss body and type 316ss sintered support screen. Back-pressure support screen available to prevent membrane rupture and deflect flow at very high pressures. Inlet /Outlet connections are 1/4" NPTF.

Applications: Use for filtration of process liquids or gases in-line.

Ordering information: Replacement and accessory parts are available. Filters are not included.



 

ORDER

Expand All / Collapse All
Material Model Filter Diameter Prefilter Diameter Supplier No. VWR Catalog Number Unit Price Quantity
Stainless Steel LS47HP 47 mm 38 mm 304900 101105-622 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Filtration Area 12.5 cm2
Max. Differential Pressure 276 psi

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Adaptor O-ring 19304905 102968-956 Each Retrieving
Back Pressure Support Screen 304920 102968-962 Pack of 1 Retrieving
Inner O-ring 19304906 102968-958 Pack of 3 Retrieving
Outer O-ring 19304910 102968-960 Pack of 3 Retrieving
Support Screen 304903 101103-274 Pack of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty