Human Recombinant Resistin (from E. coli)
Supplier: BioVision
AB285589-100UG AB285589-25UG AB285589-1MG
10006-768EA 1018.89 USD
10006-768 10006-770 10006-772
Human Recombinant Resistin (from E. coli)
Proteins and Peptides
>98% Pure Recombinant human Resistin.
Human Resistin is a new member to the family of adipocyte secreted proteins. The biological functions of Resistin as well as its molecular target is largely unknown. Whether it promotes or antagonizes insulin action is still unclear. Recombinant Resistin is a 19.5 kDa dimeric protein consisting of two 92 amino acid polypeptide chains.