Would you like to log in again?
Endoglin, human recombinant, BioVision
Supplier: BioVision
4530-10 4530-100
10006-746EA 329.6 USD InStock
10006-746 10006-748
Endoglin, human recombinant, BioVision
Proteins and Peptides
>95% Pure Recombinant human Endoglian.
Endoglin is a type I membrane glycoprotein located on cell surfaces and is part of the TGF beta receptor complex.The protein consists of a homodimer of 180 kDA with disulfide links. It has been found on endothelial cells, activated macrophages, fibroblasts, and smooth muscle cells. Endoglin has been found to be part of the TGF-beta1 receptor complex. It thus may be involved in the binding of TGF-beta1, TGF-beta3, Activin-A, BMP-2, and BMP-7. It has been postulated that Endoglin is involved in the cytoskeletal organization affecting cell morphology and migration. Endoglin has a role in the development of the cardiovascular system and in vascular remodeling. Its expression is regulated during heart development . CD105 Human Recombinant extracellular domain produced in baculovirus is a homodimeric, glycosylated, polypeptide containing 586 amino acids and having a molecular mass of 61 kDa but as a result of glycosylation, migrates at 90 kDa under reducing conditions in SDS-PAGE. The CD105 is fused to a C-terminal His-tag (6xHis) and purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty