Would you like to log in again?
Cytochrome Oxidase Activity Assay Kit, BioVision
Supplier: BioVision
K287-100
10013-672EA 464.82 USD InStock
10013-672
Cytochrome Oxidase Activity Assay Kit, BioVision
Assays Protein Assays
Rapid, HTS
  • Simple and rapid measurement of cytochrome oxidase enzymatic activity in 96-well plate format
  • Useful for mitochondrial respiration studies, assembly of the complexes, mitochondria outer membrane integrity
Cytochrome c Oxidase (EC 1.9.3.1) or Complex IV is the fourth complex of the Electron Transport Chain located in the mitochondrial (or bacterial) membrane. It provides energy to the cell by coupling electron transport through the Cytochrome c chain with the process of oxidative phosphorylation. Complex IV contains 13 different subunits encoded by both mitochondrial DNA and nuclear DNA. It receives an electron from each of the four Cytochrome c molecules, and transfers it to one oxygen molecule, converting it into two molecules of water. In this process, it also binds to four proton molecules and translocates them across the membrane to establish electrochemical gradient, which is utilized for the synthesis of ATP. Cytochrome Oxidase Activity Assay Kit is simple, fast and high-throughput adaptable. This assay kit can be used for purified mitochondria or tissue extracts containing mitochondria. The activity of the enzyme is determined colorimetrically by following the oxidation of reduced Cytochrome c as an absorbance decrease at 550 nm.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty