Caspglow Active Caspase-12 Staining Kit, BioVision
K172-25 K172-100
10013-632EA 189.37 USD
10013-632 10013-630
Caspglow Active Caspase-12 Staining Kit, BioVision
Detects activated Caspase-12 by FACS, FL or plate reader
  • Fluorescence microscopy, Flow cytometry and fluorescence plate reader (Ex. = 485 nm and Em. = 535 nm)
  • Mammalian
  • Convenient sizes (25 and 100 assays)
  • Sensitive detection of activated caspase-12 in living cells
Activation of caspases plays a central role in apoptosis. The CaspGLOW™ Fluorescein Caspase-12 Staining Kit provides a convenient means for sensitive detection of activated caspase-12 in living cells. The assay utilizes the caspase-12 inhibitor, ATAD-FMK, conjugated to FITC (FITC-ATAD-FMK) as a marker. FITC-ATAD-FMK is cell permeable, nontoxic, and irreversibly binds to activated caspase-12 in apoptotic cells.