Caspase-10 Colorimetric Assay Kit, BioVision
Supplier: BioVision
AB65663-100
10013-588EA 498.39 USD
10013-588
Caspase-10 Colorimetric Assay Kit, BioVision
Assays Protein Assays
Convenient, Highly-Sensitive Assay
  • Absorbance (400 or 405 nm)
  • Mammalian
  • Convenient sizes (25, 100, 200, 400 assays)
  • Detect early/middle stages of apoptosis; differentiate apoptosis from necrosis
Activation of ICE-family proteases/caspases initiates apoptosis in mammalian cells. The Caspase-10 Colorimetric Assay Kit provides a simple and convenient means for assaying the activity of caspases that recognize the sequence AEVD. The assay is based on spectrophotometric detection of the chromophore p-nitroanilide (pNA) after cleavage from the labeled substrate AEVD-pNA. The pNA light emission can be quantified using a spectrophotometer or a microtiter plate reader at 400- or 405 nm.