Human Recombinant CXCL14 (from E. coli)
Supplier: BioVision
AB138785-10UG
10006-488EA 467.31 USD
10006-488
Human Recombinant CXCL14 (from E. coli)
Proteins and Peptides
>95% Pure Recombinant Human CXCL14/BRAK.
The ED50 as determined by its ability to induce calcium flux of prostaglandin E2 treated THP1 human acute monocytic leukemia cells was 1.0-10.0 ng/ml.
Human Chemokine (C-X-C motif) ligand 14(hCXCL14),also known as breast and kidney-expressed chemokine (BRAK), MIP-2 gamma, kidney-expressed chemokine (KEC), and B cell and monocyte-activating chemokine (BMAC), is a CXC chemokine constitutively expressed in certain normal tissues but is reduced or absent from many established tumor cell lines and human cancers. CXCL14 is known to be a chemo-attractant for monocyte and dendritic cells. CXCL14 inhibits angiogenesis and exhibits antimicrobial activities. Mature human and mouse CXCL14 differ by only 2 residues.