Would you like to log in again?
CXCL10/IP-10/CRG-2, human recombinant, BioVision
Supplier: BioVision
4277-1000 4277-10 4277-50
10006-484 10006-482 10006-486
CXCL10/IP-10/CRG-2, human recombinant, BioVision
>95% Pure Recombinant Human CXCL10/IP-10/CRG-2.
The ED50 of rHu CXCL10 as determined by its ability to chemoattract human CXCR3 transfected BaF3 mouse proB cells was 0.02-0.06 μg/mL.
Human Chemokine (C-X-C motif) ligand 10(CXCL10),also known as Interferon gamma-inducible protein-10 (IP-10), is a non-ELR chemokine secreted by various cell types including monocytes, endothelial cells and fibroblasts in response to IFN-γ. CXCL10 functions via chemokine receptor CXCR3. CXCL10 has been attributed to several roles, such as chemo-attraction for activated T-lymphocytes, inhibition of angiogenesis and antitumor activity.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty