Would you like to log in again?
MIP-1beta, human recombinant, BioVision
Supplier: BioVision
4247-1000 4247-10
10006-432EA 4882.2 USD InStock
10006-432 10006-430
MIP-1beta, human recombinant, BioVision
Proteins and Peptides
>99% Pure active Recombinant human MIP-1beta, which plays an important role in the inflammatory response of blood monocytes and tissue microphages.
Determined by its ability to chemoattract human blood monocytes using a concentration of 5.0-20 ng/ml.
Human macrophage inflammatory protein-1 β (MIP-1 β) is a 7.6 kDa protein containing 69 amino acid residues. It plays an important role in the inflammatory response of blood monocytes and tissue macrophages.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty