Would you like to log in again?
MIG, murine recombinant, BioVision
Supplier: BioVision
4241-1000 4241-20
10006-426 10006-428
MIG, murine recombinant, BioVision
>98% Pure active Recombinant murine MIG, which acts as a chemoattractant toward monocytes, lymphocytes, and certain T cells.
The biological activity of murine MIG was determined by its ability to chemoattract human lymphocytes using a concentration range of 0.1-1.0 ng/ml.
Murine MIG (monokine induced by interferon γ) is produced by microphages and other cells. It is a member of the α chemokine family (C-X-C) of cytokines. MIG acts as a chemoattractant toward monocytes, lymphocytes, and certain T cells. Murine MIG is an 12.2 kDa protein that consists of 105 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty