Human Recombinant MIG (from E. coli)
Supplier: BioVision
4240-20 4240-1000
10006-424EA 256.86 USD
10006-424 10006-422
Human Recombinant MIG (from E. coli)
>95% Pure active Recombinant human MIG, which acts as a chemoattractant toward monocytes, lymphocytes, and certain T cells.
The ED₅₀ range = 10 -100 ng/ml, determined by the dose dependent chemotaxis with human lymphocytes cultured in the presence of IL-2.
Human MIG (monokine induced by interferon γ) is produced by microphages and other cells. It is a member of the α chemokine family (C-X-C) of cytokines. MIG acts as a chemoattractant toward monocytes, lymphocytes, and certain T cells. Human MIG is an 11.7 kDa protein that consists of 103 amino acid residues.