Would you like to log in again?
M-CSF, murine recombinant, BioVision
Supplier: BioVision
4238-10 4238-1000 4238-50
10006-416EA 201.6 USD InStock
10006-416 10006-418 10006-420
M-CSF, murine recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant murine M-CSF, which is involved in both the development and function of monocytes/microphages.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of murine M-NSF-60 cells is <1.0 ng/ml, corresponding to a specific activity of >1 x 10⁶ units/mg.
Macrophage Colony stimulating Factor (M-CSF) is produced by various cell types, including fibroblasts, microphages, and endothelial cells. M-CSF is involved in both the development and function of monocytes/microphages. The recombinant M-CSF is a disulfide-linked homodimer composed of two 18.2 kDa subunits of 156 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty