Would you like to log in again?
MCP-4, human recombinant, BioVision
Supplier: BioVision
4231-1000 4231-10
10006-414EA 4068.5 USD InStock
10006-414 10006-412
MCP-4, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant human MCP-4.
The specific activity as determined by the ability of MCP-4 to chemoattract human monocytes was achieved at 10-100 ng/ml.
Human macrophage/monocyte chemotactic protein-4 (MCP-4) is a member of the β chemokine family (C-C) of cytokines. Like eotaxin, MCP-4 is a powerful eosinophil chemoattractant. Additionally, like MCP-3, MCP-4 elicits chemoattraction of monocytes and T lymphocytes. MCP-4 is a peptide of 8.6 kDa that consists of 75 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty