Human Recombinant SDF1 beta (from E. coli)
Supplier: BioVision
4390-100 4390-10 4390-1000
10006-290EA 642.12 USD
10006-290 10006-292 10006-288
Human Recombinant SDF1 beta (from E. coli)
>98% Pure active Recombinant human SDF-1beta (CXCL12).
Determined by its ability to chemoattract human peripheral T cells activated with PHA and IL-2 using a concentration range of 20-80 ng/ml.
Stromal cell-derived factor-1 β is a stromal derived CXC chemokine. It signals through CXCR4 receptor. Recombinant Human SDF-1β is an 8.5 kDa protein containing 72 amino acid residues.