Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

IL-27 / EBI-3, murine recombinant, BioVision

Supplier: BioVision
4377-100 4377-20 4377-1000
>90% Pure Recombinant active murine EBI-3/IL-27.

Assay data for mouse recombinant EBI-3 is based upon qualitative binding to anti-EBI-3 antibody.

Epstein-Barr Virus Induced Gene-3 (EBI-3), is a secreted glycoprotein belonging to the hematopoietin receptor family and related to the p40 subunit of IL-12. It was identified by its induced expression in B-lymphocytes in response to Epstein-Barr virus infection. EBI-3 forms heterodimers with p28 to form IL-27 and with p35 to form IL-35. Both IL-27 and IL-35 have anti-inflammatory and regulatory activity. Recombinant Mouse EBI is a non-glycosylated polypeptide chain consisting of 207 amino acids with a molecular weight of 22,900 Da.




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg 4377-100 10006-050 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

1 mg 4377-1000 10006-660 Each (1mg) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

20 μg 4377-20 10006-662 Each (20µG) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Protein/Peptide Name Ebi-3
Protein Synonyms EBI3, mouse EBI3, IL-27, Epstein-Barr virus induced 3 rh-4-1BB Receptor, 4-1BB Receptor, h-4-1BB Receptor h-4-1BB Receptor, rh-4-1BB Receptor, 4-1BB Receptor
Protein/Peptide Type Recombinant
Species Mouse
Source E. coli
Purity ≥90%
Endotoxin Content <0.01 ng/μg
Molecular Weight 22.9 kDa
Formulation Lyophilized from 10 mM Sodium citrate, pH 3.
Reconstitution Instructions Reconstitute the lyophilized product with sterile H₂O at a concentration of 0.1 – 0.5 mg/ml
Protein Type Recombinant

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty
 

A new look and a renewed commitment

Built on the features & resources you know, optimized with innovation from Avantor®. Our new brand symbolizes agility and our ability to adapt to customer needs.

Learn More Start Shopping