Human Recombinant Lymphotactin (from E. coli)
Supplier: BioVision
4375-1000 4375-10
10006-040EA 221.82 USD
10006-040 10006-042
Human Recombinant Lymphotactin (from E. coli)
>98% Pure Recombinant active human Lymphotactin.
Determined by its ability to chemoattract human peripheral blood lymphocytes using a concentration range of 10-100 ng/ml.
Lymphotactin is part of the C family chemokines that exhibit chemotactic activity toward lymphocytes but not toward monocytes or neutrophils. Recombinant Human lymphotactin is a 10 kDa protein containing 92 amino acid residues.