Human Recombinant 4-1BB Ligand (from E. coli)
Supplier: BioVision
4369-100 4369-20 4369-1000
10006-646EA 694.66 USD
10006-646 10006-648 10006-650
Human Recombinant 4-1BB Ligand (from E. coli)
>98% Pure active Recombinant human 4-1BBL.
Determined by the dose-dependent stimulation of IL-8 production by human PBMC. The ED₅₀ for the effect is less than 10 ng/ml. Please note: results may vary with PBMC donors.
Human 4-1BB Ligand (4-1BBL) is a member of the emerging family of Ligands with structural homology to tumor necrosis factor. Recombinant human 4-1BBL is a 19.5 kDa protein containing 185 amino acid residues, comprising the TNF-like extra-Cellular domain of 4-1BBL.