Mouse Recombinant Leptin (from E. coli)
Supplier: BioVision
4367-02 4367-5 4367-1
10006-636EA 233.5 USD
10006-636 10006-634 10006-638
Mouse Recombinant Leptin (from E. coli)
>98% Pure active Recombinant murine Leptin.
Determined in an in vivo assay using the ob/ob mouse obesity model and NZO. Both strains of mice were treated via intraperitoneal injection once daily at a dose of 5 µg leptin/gm of body weight for 7 days. Significant effects on body weight, food consumption, and plasma glucose levels were observed to saline-treated controls.
Leptin (Anti-Obesity Protein) is the protein product of the ob (obese) gene in mice and seems to be involved in appetite control. The ob gene is expressed in adipose tissue and is thought to regulate the body’s fat stores. Mice with the ob/ob genotype also develop a form of diabetes similar to type II (non-insulin dependent) diabetes. Murine Leptin is a 16 kDa protein containing 147 amino acid residues.