Human Recombinant Leptin (from E. coli)
Supplier: BioVision
4366-02 4366-5 4366-1
10006-628EA 93.39 USD
10006-628 10006-632 10006-630
Human Recombinant Leptin (from E. coli)
>98% Pure active Recombinant human Leptin.
BioVision’s human Leptin has been shown to be biologically active in two different mouse obesity models, ob/ob and NZO. Both strains of mice were treated via intraperitoneal injection once daily at a dose of 5 mg Leptin/gm of body weight for 7 days. Significant effects on body weight, food consumption, and plasma glucose levels were observed to saline-treated controls.
Leptin (Anti-Obesity Protein) is the protein product of the ob (obese) gene in mice and seems to be involved in appetite control. The ob gene is expressed in adipose tissue and is thought to regulate the body’s fat stores. Mice with the ob/ob genotype also develop a form of diabetes similar to type II (non-insulin dependent) diabetes. Human Leptin is a 16 kDa protein containing 146 amino acid residues.