Human Recombinant MIF (from E. coli)
Supplier: BioVision
4984-1000 4984-10 4984-100
10007-368EA 1272.57 USD
10007-368 10007-370 10007-366
Human Recombinant MIF (from E. coli)
>97% Pure active Recombinant human MIF.
Human MIF biological activity was measured in an agarose micro droplet assay using human U937 cells as targets. The optimal range of activity found for human MIF in this assay was 0.5-1µg/ml.
The cytokine Macrophage migration inhibitory factor (MIF) has been identified to be secreted by the pituitary gland and the monocyte/macrophage and to play an important role in endotoxic shock. MIF has the unique property of being released from macrophages and T cells in response to physiological concentrations of glucocorticoids. The secretion of MIF is tightly regulated and decreases at high, anti-inflammatory steroid concentration. Recombinant human MIF is a single, non-glycosylated, polypeptide chain containing 115 amino acids and having a molecular mass of 12.5 kDa.