Would you like to log in again?
MCP-1 (MCAF), human recombinant, BioVision
Supplier: BioVision
4222-1000 4222-100 4222-20
10006-392EA 2832.5 USD InStock
10006-392 10006-390 10006-394
MCP-1 (MCAF), human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant Human MCP-1 (MCAF), which plays an important role in the inflammatory response of blood monocytes and tissue microphages.
The activity is determined by its ability to chemoattract human monocytes at 4 - 15 ng/ml.
Monocyte Chemoattractant Protein 1 (MCP-1), also known as CCL2, is thought to be produced by injured or infected tissues. MCP-1 signals through G protein-coupled receptors, CCR2 and CCR4, to recruit memory T cells, monocytes and dendritic cells. Recombinant mouse MCP-1 is a non-glycosylated protein, containing 76 amino acids and having a molecular mass of 8.6 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty