Mouse Recombinant Periostin (from E. coli)
Supplier: BioVision
4205-10
10006-376EA 402.79 USD
10006-376
Mouse Recombinant Periostin (from E. coli)
>95% Pure Recombinant murine Periostin/OSF-2.
Periostin was originally isolated as a osteoblast-specific factor that functions as a cell adhesion molecule for preosteoblasts and is thought to be involved in osteoblast recruitment, attachment and spreading. Additionally, periostin expression has previously been shown to be significantly increased by both transforming growth factor β1 (TGF-β1) and bone morphogenetic protein (BMP-2). Abnormal expression of periostin is also linked to angiogenesis and metastasis in epithelial tumors. BioVision’s Mouse periostin (aa 24-783) (isoform 5) is fused at the C-terminus to a FLAG®-tag.