Human Recombinant FABP7 (from E. coli)
Supplier: BioVision
4507-1000 4507-100 4507-25
10006-734EA 402.79 USD
10006-734 10006-738 10006-736
Human Recombinant FABP7 (from E. coli)
>98% Pure Recombinant human FABP7.
Test in process
FABPs have been reported to be involved in the uptake and metabolism of fatty acids, maintenance of Cellular membrane fatty acids levels, intraCellular trafficking, modulation of specific enzymes of lipid metabolic pathways, as well as in the modulation of cell growth and differentiation.