Would you like to log in again?
BD-1, human recombinant, BioVision
Supplier: BioVision
4506-20 4506-1000
10006-732EA 195.7 USD InStock
10006-732 10006-730
BD-1, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human BD-1.
Determined by its ability to chemoattract CD34+ dendritic cells using a concentration range of 0.1-1 µg/ml.
The Defensin family are highly similar in their protein sequence and are microbicidal & cytotoxic peptides made by neutrophils. β Defensin-1 (BD-1) is an antimicrobial peptide having the resistance of epithelial surfaces to microbial colonization. BD-1 has close proximity to Defensin α-1 and has been implicated in the pathogenesis of cystic fibrosis. Skin of patients having atopic dermatitis patients and mycosis fungoides (non-lesional and lesional) show lower human BD-1 mRNA expression and higher human BD-2 and human BD-3 mRNA expression. β Defensin is highly expressed by epithelial cells. BD-1 may play a role in the pathogenesis of severe sepsis. Variation in human BD-1 contributes to asthma diagnosis, with apparent gender-specific effects. Human BD-3 is a dimer, while Human BD-1 and Human BD-2 are monomeric. Recombinant human β Defensin-1 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 47 amino acids and having a molecular mass of 5 kDa.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty