Would you like to log in again?
PDGF-BB, human recombinant, BioVision
Supplier: BioVision
4488-50 4488-1000 4488-10
10006-722EA 798.25 USD InStock
10006-722 10006-720 10006-718
PDGF-BB, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human PDGF-BB .
The ED₅₀ as determined by the dose-dependent stimulation of thimidine uptake by BALB/c 3T3 cells is < 1 ng/ml, corresponding to a specific activity of > 1 x 10⁶ units/mg.
Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle and connective tissue. PDGF, which is composed of a dimer of two chains termed the A chain and B chain, can be present as AA or BB homodimers or as an AB heterodimer. Human PDGF-BB is a 24.3 kDa B chain homodimer protein formed by subunits of 109 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty