Would you like to log in again?
VEGI/TL1 human recombinant, BioVision
Supplier: BioVision
4966-10 4966-1000 4966-50
10006-966 10006-968 10006-970
VEGI/TL1 human recombinant, BioVision
>95% Pure active Recombinant human VEGI/TL1.
The EDso as determined by the dose-dependent inhibition of the proliferation of HUVEC is <5 ug/ml.
VEGI (Vascular Endothelial Growth Inhibitor) is a member of the tumor necrosis factor family. VEGI is predominantly an endothelial cell-specific gene, and recombinant VEGI is a potent inhibitor of endothelial cell proliferation, angiogenesis and tumor growth. VEGI exerts two activities of endothelial cells: early Gl arrest of G0/G1-cells responding to growth stimuli, and programmed cell death. VEGI is also able to regulate the expression of several important genes involved in angiogenesis. Human VEGF is a 21.8 kDa homodimeric protein consisting of 192 amino acid polypeptide chains.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty