Would you like to log in again?
PDGF-AA, murine recombinant, BioVision
Supplier: BioVision
4483-10 4483-1000
10006-714EA 252.35 USD InStock
10006-714 10006-716
PDGF-AA, murine recombinant, BioVision
Proteins and Peptides
>97% Pure active Recombinant murine PDGF-AA .
In progress
Recombinant Mouse Platelet Derived Growth Factor-AA is a disulfide linked dimer comprised of 2 polypeptide chains of 126 amino acids each, with a dimeric molecular weight of 28.9 kDa. Belonging to a family of PDGF’s including PDGF-AA, PDGF-BB and PDGF-AB, they play an important role as a mitogen for a number of cell types. Recombinant Mouse PDGF-AA produced in E.Coli is a non-glycosylated homodimer.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty