Human Recombinant PDGF-AA (from E. coli)
Supplier: BioVision
4482-1000 4482-50 4482-10
10006-712EA 951.51 USD
10006-712 10006-710 10006-708
Human Recombinant PDGF-AA (from E. coli)
>98% Pure active Recombinant human PDGF-AA .
The ED₅₀ as determined by the dose-dependent stimulation of thymidine uptake by Balb-c 3T3 cells is < 1 ng/ml, corresponding to a specific activity of >1 x 10⁶ units/mg.
Platelet Derived Growth Factor (PDGF) is a potent mitogen for a wide range of cell types including fibroblasts, smooth muscle and connective tissue. PDGF, which is composed of a dimer of two chains termed the A chain and B chain, can be present as AA or BB homodimers or as an AB heterodimer. Human PDGF-AA is a 28.5 kDa A chain homodimer protein containing 250 amino acid residues.