Would you like to log in again?
VEGF121, human recombinant, BioVision
Supplier: BioVision
4963-1000 4963-10 4963-50
10006-866EA 3708 USD InStock
10006-866 10006-864 10006-964
VEGF121, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human VEGF121.
Determined by the dose-dependent stimulation of the proliferation of human μmbilical vein endothelial cells (HUVEC) using a concentration range of 0.1-0.5 ng/ml.
VEGF (Vascular Endothelial Growth Factor) is a secreted homodimeric protein, secreted by a variety of Vascularized tissues. VEGF stimulates endothelial cell growth, angiogenesis, and capillary permeability. Human VEGF is a 28.4 kDa homodimeric protein consisting of two 121 amino acid polypeptide chains.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty