Would you like to log in again?
Midkine, human recombinant, BioVision
Supplier: BioVision
4479-100 4479-20 4479-1000
10006-702EA 612.85 USD InStock
10006-702 10006-706 10006-704
Midkine, human recombinant, BioVision
Proteins and Peptides
>98% Pure Recombinant active human Midkine
Determined by its ability to chemoattract human neutrophils using a concentration of 0.1-10 ng/ml.
Human Midkine (MK) is a new member of the heparin-binding neurotrophic factor family. MK and PTN (Pleiotrophin) are structural homologs, and are highly conserved among species. MK plays important roles in development and carcinogenesis and has several important biological effects, including promotion of neurite extension and neuronal survival. Recombinant midkine is a 13.4 kDa protein, comprising of 123 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty