Human Recombinant Pleiotrophin Ptn (from E. coli)
Supplier: BioVision
4469-20 4469-1000
10006-700EA 268.54 USD
10006-700 10006-698
Human Recombinant Pleiotrophin Ptn (from E. coli)
>98% Pure Recombinant human Pleiotrophin (PTN).
Human Pleiotrophin (PTN) also called HARP (Heparin affin regulatory peptide) is a new member of the heparin-binding neurotrophic factor family. PTN (Pleiotrophin) and MK (midkine) are structural homologs, and are highly conserved among species. PTN plays an important role in development processes and promotion of neurite extension. Recombinant human PTN is a His-tag fusion protein (17.3 kDa) containing 136 amino acid residues of the PTN.