Would you like to log in again?
Oncostatin M, human recombinant, BioVision
Supplier: BioVision
4466-1000 4466-10
10006-696EA 5356 USD InStock
10006-696 10006-694
Oncostatin M, human recombinant, BioVision
Proteins and Peptides
>98% Pure active Recombinant human Oncostatin M.
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of human TF-1 cells is <2 ng/ml, corresponding to a specific activity of >5 x 10⁵ units/mg.
Oncostatin M (OSM) is an important growth regulating cytokine that affects, variably, a number of tumor and normal cells. Oncostatin M exerts inhibitory effects on the growth of A375 melanoma and other cellos, but augments the growth of normal fibroblasts, AIDs-related Kaposi sarcoma cells, and certain other cells. Recombinant Human Oncostatin M is a 26 kDa protein containing 227 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty