Would you like to log in again?
BAFF, rat recombinant, BioVision
Supplier: BioVision
4453-100 4453-20 4453-1000
10006-688EA 726.74 USD InStock
10006-688 10006-692 10006-690
BAFF, rat recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant rat BAFF.
Rat BAFF (for B cell activating factor belonging to the TNF family) is a newly discovered novel ligand of the TNF family. BAFF plays an important role as co-stimulator of B cell proliferation and function. Recombinant human BAFF is a soluble protein containing 184 amino acid residues (MW 20.7 kDa).
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty