Would you like to log in again?
BAFF, human recombinant, BioVision
Supplier: BioVision
4451-1000 4451-100 4451-500 4451-20
10006-676EA 4377.5 USD InStock
10006-676 10006-298 10006-680 10006-678
BAFF, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant human BAFF.
Determined by the dose-dependent stimulation of IL-8 production by human PBMC. The ED₅₀ for this effect is less than 10 ng/ml. Please note: results may vary with PBMC donors.
Human BAFF (for B cell activating factor belonging to the TNF family) is a newly discovered novel ligand of the TNF family. BAFF plays an important role as co-stimulator of B cell proliferation and function. Recombinant human BAFF is a soluble 17.0 kDa protein containing 153 amino acid residues
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty