Would you like to log in again?
VEGFR2, human recombinant, BioVision
Supplier: BioVision
4952-100 4952-10
10006-858EA 1668.6 USD InStock
10006-858 10006-856
VEGFR2, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant human VEGFR2.
The activity of VEGFR2 D1-7 was determined by its ability to abolish the binding of iodinated VEGF to solid surfaces or cell surfaces receptors. The ED50 for this effect is typically 10.0 ng/ml, corresponding to a specific activity of 100,000IU/mg.
VEGFR-2 has a lower affinity for VEGF than the Flt-1 receptor, but a higher signaling activity. Mitogenic activity in endothelial cells is mainly mediated by VEGFR-2 leading to their proliferation. No naturally occurring, secreted forms of VEGFR-2 have so far been reported. The binding of VEGF165 to VEGFR-2 is dependent on heparin. Soluble VEGFR-2 Human Recombinant produced in baculovirus is monomeric, glycosylated, polypeptide having a molecular mass of 116 kDa. The soluble receptor protein contains only the first 7 extracellular domains, which contain all the information necessary for ligand binding. The sKDR is purified by proprietary chromatographic techniques.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty