Would you like to log in again?
SUMO2, human recombinant, BioVision
Supplier: BioVision
4942-1000 4942-100
10007-350EA 1390.5 USD InStock
10007-350 10007-348
SUMO2, human recombinant, BioVision
Proteins and Peptides
>95% Pure active Recombinant human SUMO2.
Typical in vitro concentrations for conjμgate formation is 10-50 µM depending on conditions.
SUMO modification has been implicated in functions such as nuclear transport, chromosome segregation and transcriptional regulation. SUMO1 functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. Still, unlike ubiquitin which targets proteins for degradation, SUMO-2 is involved in a variety of Cellular processes, for example nuclear transport, transcriptional regulation, apoptosis, and protein stability. The active recombinant SUMO-2 is derived from the precursor pro-SUMO-2 (Accession # NM_006937). Human SUMO-2 shares 44% and 86% identity with SUMO-1 and SUMO-3 respectively. SUMOylation can occur without the requirement of a specific E3 ligase activity, where SUMO is transferred directly from UbcH9 to specific substrates. SUMOylated substrates are primarily localized to the nucleus (RanGAP-1, RANBP2, PML, p53, Sp100, HIPK2) but there are also cytosolic substrates (IκBα, GLUT1, GLUT4). SUMO modification has been implicated in functions such as nuclear transport, chromosome segregation, transcriptional regulation, apoptosis and protein stability.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty