Human Recombinant SUMO1 (from E. coli)
Supplier: BioVision
4941-100 4941-1000
10007-346EA 1575.67 USD
10007-346 10007-344
Human Recombinant SUMO1 (from E. coli)
>90% Pure active Recombinant human SUMO1.
Typical in vitro concentrations for conjμgate formation is 10-50 µM depending on conditions.
SUMO modification has been implicated in functions such as nuclear transport, chromosome segregation and transcriptional regulation. SUMO1 functions in a manner similar to ubiquitin in that it is bound to target proteins as part of a post-translational modification system. Still, unlike ubiquitin which targets proteins for degradation, SUMO1 is involved in a variety of Cellular processes, for example nuclear transport, transcriptional regulation, apoptosis, and protein stability. SUMO1 is not active until the last four amino acids of the carboxy-terminus are cleaved off. The active recombinant SUMO-1 protein is derived from the precursor pro-SUMO-1 (Accession # NM_003352). Human SUMO-1 shares 46% and 47% identity with SUMO-2 and SUMO-3 respectively. SUMOylation can occur without the requirement of a specific E3 ligase activity, where SUMO is transferred directly from UbcH9 to specific substrates. SUMOylated substrates are primarily localized to the nucleus (RanGAP-1, RANBP2, PML, p53, Sp100, HIPK2) but there are also cytosolic substrates (IκBα, GLUT1, GLUT4).