Nitrile Gloves, 13" Flock-Lined, Tronex®
Supplier: Tronex
9655-20 9655-30 9655-35 9655-10
10818-530CS 922.2 USD
10818-530 10818-528 10818-526 10818-524
Nitrile Gloves, 13" Flock-Lined, Tronex®
Tronex® Flock lined nitrile gloves are an excellent choice for industrial grade protection, coverage, and handling.
  • 100% latex-free
  • 13" length for greater protection; 14 to 17 mil thickness
  • Hand-specific pairs (left and right) for comfort and reduced hand fatigue
  • Soft flocked lining helps absorb perspiration
  • Raised diamond texture for confident grip
  • Excellent resistance against oils, fats, bleaching agents, and solvents
This glove features flock-lined Nitrile to ensure maximum solvent and chemical resistance.