Anti-4-HNE Goat Polyclonal Antibody

Supplier: Cell Biolabs
STA-034
102966-222EA 530 USD
102966-222
Anti-4-HNE Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: 4-HNE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR