Anti-Apolipoprotein B Goat Polyclonal Antibody

STA-134
102964-808EA 473.85 USD
102964-808
Anti-Apolipoprotein B Goat Polyclonal Antibody
Type: Primary
Antigen: Apolipoprotein B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR