Anti-Apolipoprotein AI Goat Polyclonal Antibody

STA-132
102964-804EA 473.85 USD
102964-804
Anti-Apolipoprotein AI Goat Polyclonal Antibody
Type: Primary
Antigen: Apolipoprotein AI
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR