Anti-MDA Goat Polyclonal Antibody

Supplier: Cell Biolabs
STA-031
102964-796EA 395 USD
102964-796
Anti-MDA Goat Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MDA
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype:
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR