Anti-Methylglyoxal Mouse Monoclonal Antibody [clone: 3D11]

Supplier: Cell Biolabs
STA-011
102964-790EA 523.08 USD
102964-790
Anti-Methylglyoxal Mouse Monoclonal Antibody [clone: 3D11]
Antibodies
Type: Primary
Antigen: Methylglyoxal
Clonality: Monoclonal
Clone: 3D11
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR