Anti-HA Mouse Monoclonal Antibody [clone: HA.C5]

Supplier: Cell Biolabs
AKR-006
102963-594EA 395 USD
102963-594
Anti-HA Mouse Monoclonal Antibody [clone: HA.C5]
Antibodies
Type: Primary
Antigen: HA
Clonality: Monoclonal
Clone: HA.C5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG3
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR