Anti-HIS Mouse Monoclonal Antibody [clone: HIS.H8]

Supplier: Cell Biolabs
AKR-003
102963-588EA 498.46 USD
102963-588
Anti-HIS Mouse Monoclonal Antibody [clone: HIS.H8]
Antibodies
Type: Primary
Antigen: HIS
Clonality: Monoclonal
Clone: HIS.H8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Multiple
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR