Would you like to log in again?
Nitrile Gloves, 13" Premium Gauge, Tronex®
Supplier: Tronex
Tronex®
9500-35 9500-20 9500-10 9500-30
10818-522CS 582.48 USD InStock
10818-522 10818-518 10818-516 10818-520
Nitrile Gloves, 13" Premium Gauge, Tronex®
Gloves
Specially formulated nitrile compound provides excellent barrier protection and durability with a high degree of resistance to chemicals, punctures, abrasions, and tears
  • 100% latex-free, powder free, and nonsterile
  • 13" length for additional coverage and protection
  • Raised diamond texture for a secure grip
  • Hand-specific pairs (left and right) for a comfortable fit
Unlined interior surface offers enhanced tactile sensitivity, flexibility, and dexterity for reduced hand fatigue.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty