Would you like to log in again?
Mini and Midi Horizontal Electrophoresis Productivity Packages Bundles, Expedeon
Supplier: Expedeon
GCMHU-2050 GCMHU-2050PS
10790-472EA 994.94 USD InStock
10790-472 10790-470
Mini and Midi Horizontal Electrophoresis Productivity Packages Bundles, Expedeon
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
CBS Mini and Midi Horizontal Productivity Packages are ideal for separation and identification of DNA fragments, RNA fragments, PCR products and synthetic oligonucleotides
Platinum electrode cassettes provide rapid sample velocity and clear focused bands without smiling. These cassettes are easily removed for replacement or cleaning. All gel trays are winged for easy handling. No more sticking fingers into slippery buffer to extract gel tray. Mini gel trays feature etched migration ruler on base with 1mm calibrated gel anchors on all four corners and two comb slots.
Tapered baffles allow easy in situ gel formation with or without gel tray. Combs are printed with catalog number and thickness for easy identification.
Universal input power supply has a light, compact, and stackable design. Universal input power supply accepts all domestic and international voltages. Parameters are set by pressing UP/DOWN arrow buttons in combination with a MODE button. Power output is controlled by setting a maximum value for voltage or current. The power supply automatically “crosses over” or switches the limiting parameter according to the programmed limits as the resistance of the electrical load changes during a run. Safety features include no load detection, sudden load change protection, over voltage protection and automatic power failure recovery.
Great for academic and start up labs. Systems are high quality, durable heavy duty construction, featuring a 3 Year warranty with vast array of accessory options. Productivity packages allow one gel to be running while pouring the next. Mini has gel bed dimensions of 7.5cm (w) x 10cm (l) with maximum number of samples of 32. Midi has gel bed dimension of 14cm (w) x 10cm (l) with maximum number of samples of 54.
Ordering information: Mini and Midi Horizontal Productivity Packages Include: Mini and Midi Electrophoresis units with molded tapered baffles, 2 UV transparent winged gel trays per unit, UV transparent acrylic gel beds, external gel caster per size, safety covers with power leads and Platinum electrode cassettes. Mini Horizontal combs included are 1mm 10/14 well and 1mm 8/16 double sided comb. Midi Horizontal combs included are 1mm x 20 well and 1mm x 27 well. 300V power supply included.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty