Sklarlite™ Sklarcut™ Ragnell Dissecting Scissors, OR Grade, Sklar
Supplier: Sklar
23-1250
10195-276EA 331.54 USD
10195-276
Sklarlite™ Sklarcut™ Ragnell Dissecting Scissors, OR Grade, Sklar
Forceps
Sklarlite™ Sklarcut™ Ragnell Scissors, also known as Kilner/Ragnell scissors, are a lighter version of the standard Sklar scissors that feature micro-serrations on one blade
This assists in securely holding the tissue during cutting and minimizes tissue from slipping so the most accurate and atraumatic cuts can be made. A unique manufacturing process developed by Sklar shaves microns off standard pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue. Additionally, the long shanks and short jaws of the Ragnell scissors allow the user greater control with less effort during dissection. This combined with flat blades and blunt tips often make Ragnell scissors the dissection instrument of choice during longer procedures.