Would you like to log in again?
Sera-Mag™ Magnetic Carboxylate Modified Beads, GE Healthcare
Supplier: GE Healthcare - Life Sciences
24152105050350 45152105050250 24152150350 44152105050250 44152150250 45152150250 44152150350 44152105050350
10204-630 10204-658 CA10146-026 10204-664 CA10146-060 CA10146-066 CA10146-062 10204-666
Sera-Mag™ Magnetic Carboxylate Modified Beads, GE Healthcare
The Sera-Mag Magnetic Beads are our original single shell magnetite bead.
  • Single shell magnetite bead
  • Activated via standard EDC/EDAC chemistry
  • React with free amino groups
The carboxyl groups on these beads can be activated via standard EDC/EDAC chemistry to react with free amino groups on antibodies, antigens, biomolecules and even oligos, to form amide bonds. Prior to downstream applications, samples can be mixed with these beads, whereupon these biomolecules of interest are covalently attached to the beads. Downstream applications include DNA purification, affinity purification or capture through protein-protein interactions, immunoprecipitation, oligo-oligo binding or other biomolecular interactions. Hydrophilic or hydrophobic bead nomenclature based on crosslinking requirements for the encapsulation layer.
Ordering information: Store at 2 to 8°C.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty